missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Mouse anti-Human, Clone: rBL10/411, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | Bcl-10 |
|---|---|
| Clone | rBL10/411 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
Bcl-10 Monoclonal specifically detects Bcl-10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
| Bcl-10 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 8915 | |
| IgG1 κ | |
| Protein A or G purified |
| rBL10/411 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
| Human Bcl-10 recombinant protein (epitope aa122-168) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title