missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bcl-10 Monoclonal Antibody (1185CT13.2.1.2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537643

Product Code. 17177228

  • £454.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Bcl-10 Monoclonal Antibody for Western Blot, IHC (P), Flow

BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bcl-10
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O95999
BCL10
KLH conjugated synthetic peptide between 1-143 amino acids from the human region of human BCL10.
400 μL
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1185CT13.2.1.2
Unconjugated
BCL10
AI132454; B cell CLL/lymphoma 10; B cell leukemia/lymphoma 10; B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; BCL10; Bcl-10; BCL10 immune signaling adaptor; C81403; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN; caspase-recruiting domain-containing protein; cCARMEN; cE10; c-E10; CED-3/ICH-1 prodomain homologous E10-like regulator; cellular homolog of vCARMEN; Cellular-E10; CIPER; Clap; hCLAP; IMD37; Mamm; mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10; OTTHUMP00000011647; OTTHUMP00000036080; RCD; RP11-234D19.2; R-RCD1
Mouse
Protein G
RUO
8915
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.