missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Antibody (BL10/411), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33190APC
This item is not returnable.
View return policy
Description
Bcl-10 Monoclonal antibody specifically detects Bcl-10 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| Bcl-10 | |
| Monoclonal | |
| APC | |
| B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
| Human Bcl-10 recombinant protein (epitope aa122-168) (Uniprot: O95999) | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 8915 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| BL10/411 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction