missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCKDK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549761
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total BCKDK protein.
The second major step in the catabolism of the branched-chain amino acids, isoleucine, leucine, and valine, is irreversibly catalyzed by the branched-chain alpha-keto acid dehydrogenase complex (BCKD), an inner-mitochondrial enzyme complex composed of 3 catalytic components: a branched-chain alpha-keto acid decarboxylase (E1), a dihydrolipoyl transacylase (E2), and a dihydrolipoamide dehydrogenase (E3). The complex also contains 2 enzymes that regulated the state of activity of the BCKD complex: a kinase (BCKDK), and a phosphorylase. The ubiquitously expressed kinase contains 1 histidine kinase domain. Maple syrup urine disease (MSUD) is a pathology secondary to an enzyme defect in the catabolic pathway of leucine, isoleucine, and valine. Accumulation of these amino acids and their corresponding keto acids results in encephalopathy and progressive neurodegeneration in infants not treated for MSUD.
Specifications
| BCKDK | |
| Polyclonal | |
| Unconjugated | |
| BCKDK | |
| [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial; AI327402; BCKDHKIN; BCKDK; BCKDKD; BCKD-kinase; BDK; branched chain alpha-ketoacid dehydrogenase kinase; branched chain ketoacid dehydrogenase kinase; branched-chain alpha-ketoacid dehydrogenase kinase; EC 2.7.11.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10295, 12041, 29603 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14874, O55028, Q00972 | |
| BCKDK | |
| Synthesized peptide derived from Internal of human BCKD (aa 32-58). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction