missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCKDK Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33228-20ul
This item is not returnable.
View return policy
Description
BCKDK Monoclonal antibody specifically detects BCKDK in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| BCKDK | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| BCKDHKIN, branched chain alpha-ketoacid dehydrogenase kinase, branched chain ketoacid dehydrogenase kinase, EC 2.7.11, EC 2.7.11.4, mitochondrial | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BCKDK (O14874).,, Sequence:, MILASVLRSGPGGGLPLRPLLGPALALRARSTSATDTHHVEMARERSKTVTSFYNQSAIDAAAEKPSVRLTPTMMLYAGRSQDGSHLLKSARYLQQELPV | |
| 20 μL | |
| Protein Kinase | |
| 10295 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction