missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BCKDK Polyclonal antibody specifically detects BCKDK in Human samples. It is validated for Western Blot
Specifications
Specifications
| Dilution | Western Blot |
| Gene Alias | BCKDHKIN, branched chain alpha-ketoacid dehydrogenase kinase, branched chain ketoacid dehydrogenase kinase, EC 2.7.11, EC 2.7.11.4, mitochondrial |
| Host Species | Rabbit |
| Purification Method | Antigen and protein A Affinity-purified |
| Regulatory Status | RUO |
| Research Discipline | Protein Kinase |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?