missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCKDHB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92027-0.1ml
This item is not returnable.
View return policy
Description
BCKDHB Polyclonal antibody specifically detects BCKDHB in Mouse, Rat samples. It is validated for Western Blot
Specifications
| BCKDHB | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| 2-oxoisovalerate dehydrogenase beta subunit, 2-oxoisovalerate dehydrogenase subunit beta, mitochondrial, BCKDE1B, BCKDH E1-beta, branched chain alpha-ketoacid dehydrogenase E1-beta subunit, branched chain keto acid dehydrogenase E1, beta polypeptide, Branched-chain alpha-keto acid dehydrogenase E1 component beta chain, dJ279A18.1, E1B, E1b-beta subunit of the branched-chain complex, EC 1.2.4.4, FLJ17880 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 183-392 of human BCKDHB (NP_898871.1). RSPWGCVGHGALYHSQSPEAFFAHCPGIKVVIPRSPFQAKGLLLSCIEDKNPCIFFEPKILYRAAAEEVPIEPYNIPLSQAEVIQEGSDVTLVAWGTQVHVIREVASMAKEKLGVSCEVIDLRTIIPWDVDTICKSVIKTGRLLISHEAPLTGGFASEISSTVQEECFLNLEAPISRVCGYDTPFPHIFEPFYIPDKWKCYDALRKMINY | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 594 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction