missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCKDHA Recombinant Rabbit Monoclonal Antibody (HL2405)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550519
This item is not returnable.
View return policy
Description
BCKDHA Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid (BCAA) dehydrogenase complex (BCKD; EC 1.2.4.4), an inner-mitochondrial enzyme complex that catalyzes the oxidative decarboxylation of the branched-chain alpha-ketoacids derived from isoleucine, leucine, and valine. This reaction is the second major step in the catabolism of the branched-chain amino acids (Wynn et al., 1998). The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2), and a homodimeric dihydrolipoamide dehydrogenase (E3). E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
Specifications
| BCKDHA | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P12694 | |
| BCKDHA | |
| Recombinant fragment of human BCKDHA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2405 | |
| Unconjugated | |
| BCKDHA | |
| 2-oxoisovalerate dehydrogenase (lipoamide); 2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial; BCKAD E1[a]; BCKDA; BCKDE1A; BCKDH E1-alpha; BCKDHA; branched chain alpha-ketoacid dehydrogenase subunit E1 alpha; branched chain keto acid dehydrogenase E1 alpha protein; branched chain keto acid dehydrogenase E1, alpha polypeptide; branched chain keto acid dehydrogenase subunit E1, alpha polypeptide; branched chain ketoacid dehydrogenase E1, alpha polypeptide; branched-chain alpha-keto acid dehydrogenase E1 component alpha chain; E1a; MSU; MSUD1; OVD1A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 593 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction