missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress BCIP
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| CAS | 6578-06-9 |
| Solubility | DMSO : ≥ 51 mg/mL (117.61 mM) ∣DMF : 25 mg/mL (57.65 mM∣Need ultrasonic) |
| Synonym | BCIP p-toluidine salt∣ X-phosphate p-toluidine salt |
| Content And Storage | 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Quantity | 500 mg |
| Molecular Weight (g/mol) | 433.62 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 98.0% |
| Shipping Condition | Room temperature |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction