missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BCI

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
BCI is an allosteric inhibitor of dual specificity phosphatase DUSP6 and DUSP1 (EC50 values are 8.0 μM and 13.3 μM respectively). Increases P2X7 receptor expression in neuroblastoma cells, reduces cell viability of EGFR or KRAS mutant lung cancer cells, and induces apoptosis in p53-null non-small cell lung cancer cells. In an animal model of myocardial infarction, BCI improves heart function, reduces fibrosis, and reduces inflammation by inhibiting macrophage formation. Activates FGF signaling in zebrafish embryo development to increase myocardial progenitor population and heart size.
Specifications
Specifications
| Chemical Name or Material | 3-(Cyclohexylamino)-2,3-dihydro-2-(phenylmethylene)-1 H-inden-1-one |
| CAS | 15982-84-0 |
| Quantity | 50 mg |
| Target | MKP-1/DUSP1 |
| Molecular Formula | C22H23NO |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction