missing translation for 'onlineSavingsMsg'
Learn More

BCI-121, MedChemExpress

Product Code. 16498018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498018 10 mg 10mg
16463736 25 mg 25mg
16457796 100 mg 100mg
16433947 50 mg 50mg
16400488 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498018 Supplier Medchem Express Supplier No. HY21972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BCI-121 is a SMYD3 inhibitor that impairs the proliferation of cancer cell.

BCI-121 is a SMYD3 inhibitor that impairs the proliferation of cancer cell.

Specifications

Chemical Name or Material BCI-121
CAS 432529-82-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H18BrN3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (293.93 mM)
SMILES NC(C1CCN(CC(NC2=CC=C(Br)C=C2)=O)CC1)=O
Molecular Weight (g/mol) 340.22
Formula Weight 340.22
Percent Purity 99.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.