missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCCIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38567-20ul
This item is not returnable.
View return policy
Description
BCCIP Polyclonal antibody specifically detects BCCIP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| BCCIP | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| BCCIPalpha, BRCA2 and CDKN1A interacting protein, BRCA2 and CDKN1A-interacting protein, BRCA2 and Cip1/p21 interacting protein, cdk inhibitor p21 binding protein, P21- and CDK-associated protein 1, Protein TOK-1, TOK-1, TOK-1alpha, TOK1BCCIPbeta, TOK-1beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-314 of human BCCIP (NP_510868.1).,, Sequence:, MASRSKRRAVESGVPQPPDPPVQRDEEEEKEVENEDEDDDDSDKEKDEEDEVIDEEVNIEFEAYSLSDNDYDGIKKLLQQLFLKAPVNTAELTDLLIQQNHIGSVIKQTDVSEDSNDDMDEDEVFGFISLLNLTERKGTQCVEQIQELVLRFCEKNCEKSMVEQLDKFLNDTTKPVGLLLSERFINVPPQIALPMYQQLQKELAGAHRTNKPCGKCYFYLLISKTFVEAGKNNSKKKPSNKKKAALMFANAEEEFFYEKAILKFNYSVQEESDTCLGGKWSFDDVPMTPLRTVMLIPGDKMNEIMDKLKEYLSV | |
| 20 μL | |
| Cell Cycle and Replication | |
| 56647 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction