missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAT1 Polyclonal Antibody

BCAT1 Polyclonal Antibody
Brand: Invitrogen PA5100403
This item is not returnable.
View return policy
Description
BCAT1 Polyclonal Antibody for Western Blot
Specifications
| BCAT1 | |
| Polyclonal | |
| Unconjugated | |
| Bcat1 | |
| BCAT(c); Bcat1; BCATC; BCT1; branched chain amino acid transaminase 1; branched chain amino acid transaminase 1, cytosolic; branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; branched-chain-amino-acid aminotransferase, cytosolic; cytosolic branched-chain amino acid aminotransferase; Eca39; MECA39; placental protein 18; PNAS121; PP18; Protein ECA39 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12035, 29592, 586 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24288, P54687, P54690 | |
| Bcat1 | |
| A synthesized peptide derived from human BCAT1(Accession P54687), corresponding to amino acid residues S196-L246. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction