missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAT1 Mouse anti-Human, Clone: 3C8, Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £378.00
Specifications
| Antigen | BCAT1 |
|---|---|
| Clone | 3C8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263922
|
Novus Biologicals
NBP2-59430-50UL |
50 μL |
£234.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18262661
|
Novus Biologicals
NBP2-59430-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCAT1 Monoclonal specifically detects BCAT1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| BCAT1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 586 | |
| Recombinant human BCAT1 (1-386aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 3C8 | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39 | |
| BCAT1 | |
| IgG1 κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title