Learn More
BCAT1 Antibody (OTI3F5), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-70242UV
Product Code. 18197413
  • £595.00 / 0.10mL
Description
BCAT1 Monoclonal specifically detects BCAT1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BCAT1
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
BCAT1
Full length recombinant protein of human BCAT1 (NP_005495) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Canine
IgG2a
Western Blot, Immunohistochemistry
OTI3F5
Western Blot, Immunohistochemistry
BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39
Mouse
Affinity Purified
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
586.0
Store at 4°C in the dark.