missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAS2 Polyclonal Antibody

BCAS2 Polyclonal Antibody
Brand: Invitrogen PA5101470
This item is not returnable.
View return policy
Description
BCAS2 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| BCAS2 | |
| Polyclonal | |
| Unconjugated | |
| BCAS2 | |
| 6430539P16Rik; AI132645; BCAS2; breast carcinoma amplified sequence 2; breast carcinoma-amplified sequence 2; Breast carcinoma-amplified sequence 2 homolog; C76366; C80030; DAM1; DNA amplified in mammary carcinoma 1 protein; pre-mRNA-splicing factor SPF27; Snt309; SPF27; spliceosome associated protein, amplified in breast cancer; spliceosome-associated protein SPF 27 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10286, 295334, 68183 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75934, Q9D287 | |
| BCAS2 | |
| A synthesized peptide derived from human BCAS2(Accession O75934), corresponding to amino acid residues R67-Q117. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction