missing translation for 'onlineSavingsMsg'
Learn More

BCAS2 Mouse Monoclonal Antibody, Proteintech

Product Code. 30234375 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30234375 20 μL 20µL
30236410 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30234375 Supplier Proteintech Supplier No. 680031IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The BCAS2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAS2. This antibody recognizes human, mouse, rat antigen. The BCAS2 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Tekniske data

Antigen BCAS2
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
Clone 2A12G1
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols DAM1
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein G purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10286
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 28 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.