missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BCAS1 Polyclonal antibody specifically detects BCAS1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | BCAS1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | breast carcinoma amplified sequence 1, breast carcinoma-amplified sequence 1, NABC1AIBC1Amplified and overexpressed in breast cancer, Novel amplified in breast cancer 1 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human BCAS1. The immunogen is located within amino acids 460 - 510 of BCAS1. Amino Acid Squence: DSTEKTITPPEPEPTG |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?