missing translation for 'onlineSavingsMsg'
Learn More

BCAR3 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18412316 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412316 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412316 Supplier Novus Biologicals Supplier No. NBP281789IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BCAR3 Polyclonal antibody specifically detects BCAR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAR3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias breast cancer estrogen resistance 3, dJ1033H22.2 (breast cancer estrogen resistance 3), KIAA0554, Novel SH2-containing protein 2, NSP2SH2D3Bbreast cancer estrogen resistance protein 3, SH2 domain-containing protein 3B
Host Species Rabbit
Immunogen BCAR3 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BCAR3. The immunogen is located within amino acids 30 - 80 of BCAR3. Amino Acid Squence: HGTLPRKKKGPPPIRS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8412
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.