missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAR3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81789AF488
This item is not returnable.
View return policy
Description
BCAR3 Polyclonal antibody specifically detects BCAR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| BCAR3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| breast cancer estrogen resistance 3, dJ1033H22.2 (breast cancer estrogen resistance 3), KIAA0554, Novel SH2-containing protein 2, NSP2SH2D3Bbreast cancer estrogen resistance protein 3, SH2 domain-containing protein 3B | |
| BCAR3 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BCAR3. The immunogen is located within amino acids 30 - 80 of BCAR3. Amino Acid Squence: HGTLPRKKKGPPPIRS | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8412 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction