missing translation for 'onlineSavingsMsg'
Learn More

BCAR3 Antibody (3G4), Novus Biologicals™

Product Code. 18350499 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350499 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350499 Supplier Novus Biologicals Supplier No. H00008412M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCAR3 Monoclonal antibody specifically detects BCAR3 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAR3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3G4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH39895
Gene Alias breast cancer estrogen resistance 3, dJ1033H22.2 (breast cancer estrogen resistance 3), KIAA0554, Novel SH2-containing protein 2, NSP2SH2D3Bbreast cancer estrogen resistance protein 3, SH2 domain-containing protein 3B
Host Species Mouse
Immunogen BCAR3 (AAH39895, 266 a.a. ~ 373 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YGTSPGQAREGSLTKGRPDVAKRLSLTMGGVQAREQNLPRGNLLRNKEKSGSQPACLDHMQDRRALSLKAHQSESYLPIGCKLPPQSSGVDTSPCPNSPVFRTGSEPA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8412
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.