missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAP/PIK3AP1 Antibody (OTI7A11), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72368PECY55
This item is not returnable.
View return policy
Description
BCAP/PIK3AP1 Monoclonal specifically detects BCAP/PIK3AP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| BCAP/PIK3AP1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| PIK3AP1 | |
| Full length recombinant protein of human BCAP/PIK3AP1 (NP_689552) produced in HEK293T cells. | |
| 0.1 mL | |
| 118788.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI7A11 | |
| Flow Cytometry | |
| 1810044J04Rik, B cell adaptor protein, BCAPB-cell adapter for phosphoinositide 3-kinase, B-cell phosphoinositide 3-kinase adapter protein 1, FLJ35564, phosphoinositide 3-kinase adapter protein 1, phosphoinositide-3-kinase adaptor protein 1, RP11-34E5.3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction