missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580374
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BCAM.
Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| BCAM | |
| Polyclonal | |
| Unconjugated | |
| BCAM | |
| 1200005K12Rik; antigen identified by monoclonal antibody F8; AU; Auberger B antigen; basal cell adhesion molecule; basal cell adhesion molecule (Lu and Au blood groups); basal cell adhesion molecule (Lutheran blood group); basal cell adhesion molecule Lutheran blood group; BCAM; B-CAM; B-CAM cell surface glycoprotein; B-cell adhesion molecule; CD239; F8/G253 antigen; glycoprotein 95kDa; Gplu; LU; lutheran antigen; Lutheran blood group; Lutheran blood group (Auberger b antigen included); Lutheran blood group glycoprotein; Lutheran blood group variant LUGA; lutheran glycoprotein; MSK19 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 4059 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P50895 | |
| BCAM | |
| Recombinant Human BCAM protein (Met1-Ala547). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction