missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAM (CD239) Mouse anti-Human, BUV615, Clone: B64, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751041
This item is not returnable.
View return policy
Description
BCAM (CD239) Monoclonal antibody specifically detects BCAM (CD239) in Human samples. It is validated for Flow Cytometry
Specifications
| BCAM (CD239) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| B64 | |
| Brilliant Ultraviolet 615 | |
| B-CAM; BCAM; Lutheran glycoprotein; MSK19 | |
| Human BCAM Transfected Cell Line | |
| RUO | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction