missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAM/CD239 Antibody (BRIC221), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66523AF750
This item is not returnable.
View return policy
Description
BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| BCAM/CD239 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BCAM | |
| Human erythrocytes | |
| 0.1 mL | |
| 4059.0 | |
| Human, Pig | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| BRIC221 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| NB100-66523 recognizes a non-polymorphic determinant on both the 85 and 78kD Lutheran (Lu) glycoproteins. BRIC 221 specifically recognizes an epitope in the fourth extracellular domain of Lu glycoprotein. Lutheran glycoprotein is a member of the immunoglobulin superfamily and has recently been designated CD239 (B-CAM) at the 7th leucocyte typing workshop. CD239 is expressed by erythrocytes in the peripheral blood. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction