missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress BCA

Product Code. 16459447
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459447 5 g 5g
16441569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16459447 Supplier MedChemExpress Supplier No. HY15908
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BCA is 2,2-Biquinoline-4,4-dicarboxylic acid disodium salt; Determination of Cu and protein assay.

Specifications

Quantity 5 g
Shipping Condition Room temperature
Storage Requirements 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Synonym Disodium bicinchoninate
CAS 979-88-4
Molecular Weight (g/mol) 388.28 g/mol
Solubility H2O : 100 mg/mL (257.55 mM; Need ultrasonic) ∣ DMSO : 15.3 mg/mL (39.40 mM; Need ultrasonic and warming)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.