missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BC 11 Hydrobromide, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Carbamimidothioic Acid (4-Boronophenyl)methyl Ester Hydrobromide |
| CAS | 443776-49-6 |
| Molecular Formula | C8H11BN2O2S . HBr |
| Quantity | 100 mg |
| Synonym | BC 11 Hydrobromide, Carbamimidothioic Acid (4-Boronophenyl)methyl Ester Hydrobromide |
| InChI Formula | InChI=1S/C8H11BN2O2S.BrH/c10-8(11)14-5-6-1-3-7(4-2-6)9(12)13;/h1-4,12-13H,5H2,(H3,10,11);1H |
| SMILES | Br.NC(=N)SCc1ccc(cc1)B(O)O |
| IUPAC Name | [4-(carbamimidoylsulfanylmethyl)phenyl]boronic acid;hydrobromide |
| Molecular Weight (g/mol) | 290.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction