missing translation for 'onlineSavingsMsg'
Learn More

BC 11 Hydrobromide, TRC

Product Code. 30343742
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30343742

Brand: Toronto Research Chemicals TRCB12934810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbamimidothioic Acid (4-Boronophenyl)methyl Ester Hydrobromide
CAS 443776-49-6
Molecular Formula C8H11BN2O2S . HBr
Quantity 10 mg
Synonym BC 11 Hydrobromide, Carbamimidothioic Acid (4-Boronophenyl)methyl Ester Hydrobromide
InChI Formula InChI=1S/C8H11BN2O2S.BrH/c10-8(11)14-5-6-1-3-7(4-2-6)9(12)13;/h1-4,12-13H,5H2,(H3,10,11);1H
SMILES Br.NC(=N)SCc1ccc(cc1)B(O)O
IUPAC Name [4-(carbamimidoylsulfanylmethyl)phenyl]boronic acid;hydrobromide
Molecular Weight (g/mol) 290.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.