missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568584
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 100%.
BBX gene ontology annotations related to this gene include DNA binding.
Specifications
| BBX | |
| Polyclonal | |
| Unconjugated | |
| BBX | |
| 5530401J07Rik; 5730403O13Rik; Ag recognized by Treg cells 1; ARTC1; BBX; BBX high mobility group box domain containing; BBX, HMG-box containing; bobby sox homolog; bobby sox homolog (Drosophila); HBP2; HMG box transcription factor BBX; HMG box-containing protein 2; HMG-BOX transcription factor BBX; HSPC339; MDS001; x 001 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56987 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WY36 | |
| BBX | |
| synthetic peptide directed towards the N-terminal region of human BBX. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction