Learn More
Invitrogen™ BBS9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114245
Description
Immunogen sequence: TDSFLTVSSC QQVESYKYQV LAFATDADKR QETEQQKLGS GKRLVVDWTL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 93%.
The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. Required for proper BBSome complex assembly and its ciliary localization.
Specifications
| BBS9 | |
| Polyclonal | |
| Unconjugated | |
| BBS9 | |
| B1; Bardet-Biedl syndrome 9; Bardet-Biedl syndrome 9 (human); bardet-Biedl syndrome 9 protein; Bardet-Biedl syndrome 9 protein homolog; BBS9; C18; D1; E130103I17Rik; EST 3159894; parathyroid hormone-responsive B1 gene protein; Parathyroid hormone-responsive B1 gene protein homolog; protein PTHB1; PTHB1; PTH-responsive osteosarcoma B1 protein; RGD1308472 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3SYG4 | |
| BBS9 | |
| Synthetic peptide directed towards the N-terminal region of Human BBS9 (aa 190-239). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.