missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBS7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106343
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BBS7.
BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.
Specifications
| BBS7 | |
| Polyclonal | |
| Unconjugated | |
| BBS7 | |
| 8430406N16Rik; Bardet-Biedl syndrome 7; Bardet-Biedl syndrome 7 (human); Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; Bbs2l1; BBS2-like 1; BBS2-like protein 1; BBS7; FLJ10715 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361930, 55212, 71492 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IWZ6, Q8K2G4 | |
| BBS7 | |
| A synthesized peptide derived from human BBS7(Accession Q8IWZ6), corresponding to amino acid residues E399-A449. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction