missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBS7 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537952
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.
Specifications
| BBS7 | |
| Polyclonal | |
| Unconjugated | |
| BBS7 | |
| 8430406N16Rik; Bardet-Biedl syndrome 7; Bardet-Biedl syndrome 7 (human); Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; Bbs2l1; BBS2-like 1; BBS2-like protein 1; BBS7; FLJ10715 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 361930, 55212 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8IWZ6 | |
| BBS7 | |
| Peptide with sequence HCILEEADHLQE, from the internal region of BBS7 (aa 647-658). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction