missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13078-1-AP-150UL
This item is not returnable.
View return policy
Description
Probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. May play a role in protein processing in limb, cardiac and reproductive system development. May play a role in cytokinesis.Specifications
| BBS6 | |
| Polyclonal | |
| Unconjugated | |
| MKKS | |
| BBS6, HMCS, KMS, McKusick Kaufman syndrome, MKKS, MKS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311456, 59030, 8195 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JI70, Q9NPJ1 | |
| Mkks | |
| BBS6 Fusion Protein Ag3785 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction