missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBS4 Monoclonal Antibody (1292CT845.130.218)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537638
This item is not returnable.
View return policy
Description
BBS4 Monoclonal Antibody for Western Blot, IHC (P)
BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.
Specifications
| BBS4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8C1Z7, Q96RK4 | |
| BBS4 | |
| recombinant protein between 1-240 amino acids from human BBS4. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1292CT845.130.218 | |
| Unconjugated | |
| BBS4 | |
| AW537059; AW742241; Bardet-Biedl syndrome 4; Bardet-Biedl syndrome 4 (human); Bardet-Biedl syndrome 4 homolog; Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein homolog; BBS4; D9Ertd464e; zgc:152964 | |
| Mouse | |
| Protein G | |
| RUO | |
| 102774, 300754, 585 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction