missing translation for 'onlineSavingsMsg'
Learn More

BBS2 Mouse anti-Canine, Human, Clone: 2E11B3, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen BBS2
Clone 2E11B3
Concentration 1.32 mg/mL
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16847945
View Documents
Proteintech
66246-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16837945
View Documents
Proteintech
66246-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by severe pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene forms a multiprotein BBSome complex with six other BBS proteins.
Specifications

Specifications

BBS2
1.32 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Bardet Biedl syndrome 2, BBS, BBS2
BBS2
IgG3
Protein A
Antibody
2E11B3
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Mouse
Canine, Human
Q9BXC9
478121, 583
BBS2 Fusion Protein Ag21390
Primary
-20°C
BBS2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.