missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06335-50ul
This item is not returnable.
View return policy
Description
BBS2 Polyclonal antibody specifically detects BBS2 in Human samples. It is validated for Western Blot
Specifications
| BBS2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Bardet-Biedl syndrome 2, Bardet-Biedl syndrome 2 protein, BBS, MGC20703 | |
| Produced in rabbits immunized with E. coli-derived Human BBS2 fragment. | |
| 50 μg | |
| Vision | |
| 583 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction