missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113189
This item is not returnable.
View return policy
Description
BBS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development.
Specifications
| BBS1 | |
| Polyclonal | |
| Unconjugated | |
| BBS1 | |
| AI451249; Bardet-Biedl syndrome 1; Bardet-Biedl syndrome 1 (human); Bardet-Biedl syndrome 1 homolog; Bardet-Biedl syndrome 1 protein; Bardet-Biedl syndrome 1 protein homolog; BBS1; BBS2L2; BBS2-like protein 2; D19Ertd609e | |
| Rabbit | |
| Protein G | |
| RUO | |
| 52028, 582 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3V3N7, Q8NFJ9 | |
| BBS1 | |
| Recombinant Human Bardet-Biedl syndrome 1 protein (217-417AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction