Learn More
Brady™ BBP11 Monochrome UK Printer
Product Code. 11583235
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
11583235 Each
1 options
Product Code. 11583235 Supplier Brady™ Supplier No. 361075
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Brady™ BBP11 printer is a small and compact desktop printer which provides high quality thermal transfer printing at a low cost. The printer offers high performance with durable and reliable output, with a printout speed of up to 127mm (5inch) per second. It is easy to use and offers quick ribbon and media loading. The standard internal Ethernet port enables easy integration into any application.
  • Small and compact only 101.6mm (4inch) wide
  • 300dpi print resolution
  • Offers a wide range of printable materials
  • Reliable and durable
  • Many features built-in as standard such as Ethernet (network), triple communication interface and dual walled construction Ideal entry level thermal transfer printer for blood, tissue and slide labels in hospitals, healthcare and medical laboratories.
Specifications
Depth (Metric) 258 mm