missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBOX1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97720PCP
This item is not returnable.
View return policy
Description
BBOX1 Polyclonal antibody specifically detects BBOX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BBOX1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 2-oxoglutarate dioxygenase, 2-oxoglutarate dioxygenase 1, BBHgamma-butyrobetaine, BBOXEC 1.14.11.1, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetainehydroxylase) 1, Gamma-BBH, Gamma-butyrobetaine, gamma-butyrobetaine dioxygenase, Gamma-butyrobetaine hydroxylase, G-BBH | |
| Produced in rabbits immunized with E. coli-derived Human BBOX1 fragment. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 8424 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction