missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBOX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | BBOX1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661492
|
Novus Biologicals
NBP2-97720-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680795
|
Novus Biologicals
NBP2-97720-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BBOX1 Polyclonal antibody specifically detects BBOX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| BBOX1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| PBS (pH 7.0) | |
| 8424 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 2-oxoglutarate dioxygenase, 2-oxoglutarate dioxygenase 1, BBHgamma-butyrobetaine, BBOXEC 1.14.11.1, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetainehydroxylase) 1, Gamma-BBH, Gamma-butyrobetaine, gamma-butyrobetaine dioxygenase, Gamma-butyrobetaine hydroxylase, G-BBH | |
| Produced in rabbits immunized with E. coli-derived Human BBOX1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title