Learn More
BD BBL™ Taxo™ Blank Discs
Product Code. 11762914
Change view
Click to view available options
Diameter (English) Outside:
1/4 in.
Unit Size:
Pack of 300
Product Code. Diameter (English) Outside unitSize
11762914 1/4 in. Pack of 300
1 options
Product Code. 11762914 Supplier BD Supplier No. 231039
Provides a cost effective identification and differentiation tool. BD™ BBL™ Taxo™ Blank Discs are easy-to-use discs for classical identification of microorganisms. The Taxo product line includes discs for identifying a variety of organisms.
Order Info
50 discs per vial; six vials per pack
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Identification and differentiation of microorganisms
For Use With (Equipment) Taxo™ brand testing disc
Packaging Type BBL Blank Test Discs
Quantity 6 Pack
Diameter (English) Outside 1/4 in.
Diameter (Metric) Outside 6.35 mm