missing translation for 'onlineSavingsMsg'
Learn More

Bazedoxifene acetate, MedChemExpress

Product Code. 16480708
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480708

Brand: Medchem Express HYA0036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bazedoxifene acetate (TSE-424 acetate) is an oral, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. Bazedoxifene acetate can be used for the research of osteoporosis. Bazedoxifene acetate also acts as an inhibitor of IL-6/GP130 protein-protein interactions and can be used for the research of pancreatic cancer.

Bazedoxifene acetate (TSE-424 acetate) is an oral, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. Bazedoxifene acetate can be used for the research of osteoporosis. Bazedoxifene acetate also acts as an inhibitor of IL-6/GP130 protein-protein interactions and can be used for the research of pancreatic cancer.

Specifications

Chemical Name or Material Bazedoxifene acetate
CAS 198481-33-3
Color White
Purity Grade Notes Research
Molecular Formula C32H38N2O5
Quantity 5 mg
Synonym TSE-424 acetate
Solubility Information DMSO : ≥ 100 mg/mL (188.45 mM) ∣H2O : 1 mg/mL (1.88 mM; Need ultrasonic)
SMILES CC1=C(C2=CC=C(O)C=C2)N(CC3=CC=C(OCCN4CCCCCC4)C=C3)C5=CC=C(O)C=C51.CC(O)=O
Molecular Weight (g/mol) 530.65
Formula Weight 530.65
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.