missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAZ2B, Mouse anti-Human, Clone: 2D10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00029994-M07.100ug
This item is not returnable.
View return policy
Description
Sequence: GNKDLKKRKMEENTSINLSKQESFTSVKKPKRDDSKDLALCSMILTEMETHEDAWPFLLPVNLKLVPGYKKVIKKPMDFSTIREKLSSGQYPNLETFALSpecifications
| BAZ2B | |
| Monoclonal | |
| Unconjugated | |
| bromodomain adjacent to zinc finger domain, 2B | |
| DKFZp434H071/DKFZp762I0516/FLJ45644/WALp4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2D10 | |
| In 1x PBS, pH 7.4 | |
| NM_013450 | |
| BAZ2B | |
| BAZ2B (NP_038478.1, 1833 a.a. ∼ 1931 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 29994 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction