missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAZ2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35768-100ul
This item is not returnable.
View return policy
Description
BAZ2A Polyclonal antibody specifically detects BAZ2A in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| BAZ2A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| bromodomain adjacent to zinc finger domain protein 2A, bromodomain adjacent to zinc finger domain, 2A, DKFZp781B109, FLJ45876, hWALp3, KIAA0314FLJ13768, Tip5, TIP5FLJ13780, Transcription termination factor I-interacting protein 5, TTF-I interacting peptide 5, TTF-I-interacting protein 5, WALp3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 97-295 of human BAZ2A (TIP5) (NP_038477.2).,, Sequence:, QYPSANPGSNLKDPPLLSQFSGGQYPLNGILGGSRQPSSPSHNTNLRAGSQEFWANGTQSPMGLNFDSQELYDSFPDQNFEVMPNGPPSFFTSPQTSPMLGSSIQTFAPSQEVGSGIHPDEAAEKEMTSVVAENGTGLVGSLELEEEQPELKMCGYNGSVPSVESLHQEVSVLVPDPTVSCLDDPSHLPDQLEDTPILS | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11176 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction