missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAZ1B Recombinant Rabbit Monoclonal Antibody (23GB3685), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | BAZ1B |
|---|---|
| Clone | 23GB3685 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088634
|
Invitrogen™
MA553634 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088262
|
Invitrogen™
MA553635 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAZ1B Recombinant Monoclonal antibody specifically detects BAZ1B in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| BAZ1B | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9UIG0, Q9Z277 | |
| 22385, 368002, 9031 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3685 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BAZ1B; bromodomain adjacent to zinc finger domain 1B; bromodomain adjacent to zinc finger domain protein 1B; bromodomain adjacent to zinc finger domain, 1B; bromodomain adjacent to zinc finger domain, 1B; Williams-Beuren syndrome chromosome region 9 homolog; C87820; hWALp2; transcription factor WSTF; tyrosine-protein kinase BAZ1B; tyrosine-protein kinase BAZ1B; LOW QUALITY PROTEIN: tyrosine-protein kinase BAZ1B; Tyrosine-protein kinase BAZ1B-like protein; WBSC10; WBSCR10; WBSCR9; williams syndrome transcription factor; Williams syndrome transcription factor homolog; williams-Beuren syndrome chromosomal region 10 protein; Williams-Beuren syndrome chromosomal region 9 protein; williams-Beuren syndrome chromosomal region 9 protein homolog; Williams-Beuren syndrome chromosome region 9 homolog; Wstf | |
| A synthetic peptide of human WSTF (1-50AA). | |
| Primary | |
| -20°C | |
| BAZ1B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title