missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAZ1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540610
This item is not returnable.
View return policy
Description
Peptide sequence: SLPKRGRPQV RLPVKTRGKL SSSFSSRGQQ QEPGRYPSRS QQSTPKTTVS Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%.
BAZ1A may play a role in transcriptional regulation. May be involved in the formation or maintenance of heterochromatin playing a critical role in developmental control.
Specifications
| BAZ1A | |
| Polyclonal | |
| Unconjugated | |
| BAZ1A | |
| ACF1; ATP-dependent chromatin remodeling protein; ATP-dependent chromatin-remodeling protein; ATP-utilizing chromatin assembly and remodeling factor 1; B930060C03; Baz1a; BC065123; bromodomain adjacent to zinc finger domain 1A; bromodomain adjacent to zinc finger domain protein 1A; bromodomain adjacent to zinc finger domain, 1A; cbp146; CHRAC subunit ACF1; gene trap locus 5; Gtl5; hACF1; HSPC317; hWALp1; WALp1; WCRF180; Williams syndrome transcription factor-related chromatin-remodeling factor 180 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11177 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NRL2 | |
| BAZ1A | |
| Synthetic peptide directed towards the middle region of human BAZ1A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction