Learn More
Invitrogen™ BAZ1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597272
Description
Immunogen sequence: VFVELLSPRR KRRGRKSANN TPENSPNFPN FRVIATKSSE QSRSVNIASK LSLQESESKR RCRKRQSPEP SPVTLGRRSS GRQGGVHELS AFEQLVVELV RHDDSWPFLK LVSKIQVPDY YDIIKKPIAL NIIREKVNKC EYKLASEFID DIELMFSNCF EYNPRNTSEA KAGTRLQAFF HIQAQKLGLH VTPSNVDQVS TPPAAKKSRI; Positive Samples: HeLa, Mouse lung, Mouse spleen, Rat lung, Rat skeletal muscle; Cellular Location: Nucleus.
BAZ1A may play a role in transcriptional regulation. May be involved in the formation or maintenance of heterochromatin playing a critical role in developmental control.
Specifications
| BAZ1A | |
| Polyclonal | |
| Unconjugated | |
| BAZ1A | |
| ACF1; ATP-dependent chromatin remodeling protein; ATP-dependent chromatin-remodeling protein; ATP-utilizing chromatin assembly and remodeling factor 1; B930060C03; Baz1a; BC065123; bromodomain adjacent to zinc finger domain 1A; bromodomain adjacent to zinc finger domain protein 1A; bromodomain adjacent to zinc finger domain, 1A; cbp146; CHRAC subunit ACF1; gene trap locus 5; Gtl5; hACF1; HSPC317; hWALp1; WALp1; WCRF180; Williams syndrome transcription factor-related chromatin-remodeling factor 180 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11177, 217578, 314126 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88379, Q9NRL2 | |
| BAZ1A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1347-1556 of human BAZ1A (NP_0384762). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.