Learn More
Bayonet 1140 A2B2E2K2HgP3 R D Combination Filter
Specifications
Specifications
| Certifications/Compliance | EN12941 and EN12942 |
| Filter Type | 1140 A2B2E2K2HgP3 R D |
| Product Type | Combination Filter |
| For Use With (Application) | Can be Used as Filter TH/m3 A2B2E2K2 Hg PSL in the Powered Air Purifying Respirator Dräger X-plore™ |
| Quantity | 1 Ea. |