missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAY885, MedChemExpress
Description
BAY885 is a highly potent and selective ERK5 inhibitor with an IC50 of 35 nM. BAY885 shows weak inhibition on others kinases.
Specifications
Specifications
| Chemical Name or Material | BAY885 |
| CAS | 2307249-33-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H28F3N7O2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 16.67 mg/mL (32.34 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC=C(OC(F)(F)F)C=C1N)N(CC2)CCC2C3=NC=NC4=CC(N5CCN(C)CC5)=CN=C43 |
| Molecular Weight (g/mol) | 515.53 |
| Formula Weight | 515.53 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?