missing translation for 'onlineSavingsMsg'
Learn More

Erdosteine, MedChemExpress

Product Code. 16410868
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16410868

Brand: Medchem Express HYB0289

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Erdosteine inhibits lipopolysaccharide (LPS)-induced NF-κB activation. Erdosteine has muco-modulatory, anti-bacterial, anti-inflammatory and anti-oxidant effects.

Erdosteine inhibits lipopolysaccharide (LPS)-induced NF-κB activation. Erdosteine has muco-modulatory, anti-bacterial, anti-inflammatory and anti-oxidant effects.

Specifications

Chemical Name or Material Erdosteine
Physical Form Solid
CAS 84611-23-4
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H11NO4S2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym RV 144
Solubility Information DMSO : 50 mg/mL (200.55 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (26.75 mM; Need ultrasonic)
SMILES O=C(O)CSCC(NC(CCS1)C1=O)=O
Molecular Weight (g/mol) 249.31
Formula Weight 249.31
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.